COM448650247 Combi-Blocks, Inc.

2'-Nitrophenyl-2-acetamido-2-deoxy-alpha-d-glucopyranoside

  • MDL number MFCD00006587
  • Cas Number : 10139-01-2
  • Molecular Weight : 342.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6352

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6352
Availability details for
Quantity
  • Availability
  • Add to Cart