COM448650323 Combi-Blocks, Inc.

2,7-Dimethylquinoline

  • MDL number MFCD00006763
  • Cas Number : 93-37-8
  • Molecular Weight : 157.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6430

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6430
Availability details for
Quantity
  • Availability
  • Add to Cart