COM448650370 Combi-Blocks, Inc.

4-[3-(Methoxycarbonyl)-5-isoxazolyl]benzoic acid

  • MDL number MFCD22192331
  • Cas Number : 1375064-52-0
  • Molecular Weight : 247.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6479
Availability details for
Quantity
  • Availability
  • Add to Cart