COM448650425 Combi-Blocks, Inc.

4-Methylthiopiperidine hydrochloride

  • MDL number MFCD13188535
  • Cas Number : 208245-70-9
  • Molecular Weight : 167.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6539
Availability details for
Quantity
  • Availability
  • Add to Cart