COM448650438 Combi-Blocks, Inc.

6-Chloro-1h-indole-5-carbonitrile

  • MDL number MFCD23706334
  • Cas Number : 1423120-66-4
  • Molecular Weight : 176.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6554

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6554
Availability details for
Quantity
  • Availability
  • Add to Cart