COM448650460 Combi-Blocks, Inc.

2,2'-Dichloro-1,1'-biphenyl-4,4'-diamine

  • MDL number MFCD00209459
  • Cas Number : 84-68-4
  • Molecular Weight : 253.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6582

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6582
Availability details for
Quantity
  • Availability
  • Add to Cart