COM448650570 Combi-Blocks, Inc.

Ethyl 1-cbz-3-isopropylpiperidine-3-carboxylate

  • MDL number MFCD21641318
  • Cas Number : 1363166-26-0
  • Molecular Weight : 333.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6702

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6702
Availability details for
Quantity
  • Availability
  • Add to Cart