COM448650749 Combi-Blocks, Inc.

Alpha-(chloromethyl)-2,4-dichlorobenzyl alcohol

  • MDL number MFCD00000950
  • Cas Number : 13692-14-3
  • Molecular Weight : 225.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6900

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6900
Availability details for
Quantity
  • Availability
  • Add to Cart