COM448650795 Combi-Blocks, Inc.

N-Piperidin-4-yl-benzamide

  • MDL number MFCD01314226
  • Cas Number : 33953-37-6
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6950

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-6950
Availability details for
Quantity
  • Availability
  • Add to Cart