COM448650885 Combi-Blocks, Inc.

4-Methylbenzimidazole hydrochloride

  • MDL number MFCD26404520
  • Cas Number : 1456821-64-9
  • Molecular Weight : 168.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7047

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7047
Availability details for
Quantity
  • Availability
  • Add to Cart