COM448651354 Combi-Blocks, Inc.

D-Glucoheptose

  • MDL number MFCD00006968
  • Cas Number : 62475-58-5
  • Molecular Weight : 210.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7542

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7542
Availability details for
Quantity
  • Availability
  • Add to Cart