COM448651380 Combi-Blocks, Inc.

2-Chloro-4,5-dimethyl-6-nitrophenol

  • MDL number MFCD01313712
  • Cas Number : 74098-29-6
  • Molecular Weight : 201.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7570

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7570
Availability details for
Quantity
  • Availability
  • Add to Cart