COM448651577 Combi-Blocks, Inc.

Ethyl 4-(1h-pyrrol-1-yl)benzoate

  • MDL number MFCD01313323
  • Cas Number : 5044-37-1
  • Molecular Weight : 215.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7781

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-7781
Availability details for
Quantity
  • Availability
  • Add to Cart