COM448651809 Combi-Blocks, Inc.

(3S,4S)-3-Amino-1-hydroxy-4-methyl-2-pyrrolidinone acetate

  • MDL number MFCD23381113
  • Cas Number : 1820583-82-1
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8021

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8021
Availability details for
Quantity
  • Availability
  • Add to Cart