COM448651870 Combi-Blocks, Inc.

Beta-d-ribopyranose 1,2,3,4-tetraacetate

  • MDL number MFCD00006596
  • Cas Number : 4049-34-7
  • Molecular Weight : 318.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8087

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8087
Availability details for
Quantity
  • Availability
  • Add to Cart