COM448652195 Combi-Blocks, Inc.

3-(2-Methoxyethyl)benzylamine

  • MDL number MFCD26383904
  • Cas Number : 1061650-42-7
  • Molecular Weight : 165.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8432

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8432
Availability details for
Quantity
  • Availability
  • Add to Cart