COM448652298 Combi-Blocks, Inc.

1-(4-Tetrahydropyranyl)-1,4,5,6-tetrahydropyrrolo[3,4-c]pyrazole hydrochloride

  • MDL number MFCD24335243
  • Cas Number : 1434128-47-8
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8548

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8548
Availability details for
Quantity
  • Availability
  • Add to Cart