COM448652314 Combi-Blocks, Inc.

2,4-Dibenzyloxybenzyl alcohol

  • MDL number MFCD08456666
  • Cas Number : 33617-58-2
  • Molecular Weight : 320.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8566

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8566
Availability details for
Quantity
  • Availability
  • Add to Cart