COM448652434 Combi-Blocks, Inc.

2-Bromo-4,5-di(trifluoromethyl)aniline

  • MDL number MFCD00729098
  • Cas Number : 230295-15-5
  • Molecular Weight : 308.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8695

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8695
Availability details for
Quantity
  • Availability
  • Add to Cart