COM448652452 Combi-Blocks, Inc.

5-Isopropyl-1h-pyrazole

  • MDL number MFCD16556133
  • Cas Number : 49633-25-2
  • Molecular Weight : 110.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8713

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8713
Availability details for
Quantity
  • Availability
  • Add to Cart