COM448652488 Combi-Blocks, Inc.

4-[(Methylamino)methyl]pyrocatechol hydrobromide

  • MDL number MFCD26045226
  • Cas Number : 1025423-95-3
  • Molecular Weight : 234.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8753

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-8753
Availability details for
Quantity
  • Availability
  • Add to Cart