COM448652721 Combi-Blocks, Inc.

4-Methoxy-benzenesulfonic acid

  • MDL number MFCD08685124
  • Cas Number : 5857-42-1
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9009

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9009
Availability details for
Quantity
  • Availability
  • Add to Cart