COM448652810 Combi-Blocks, Inc.

2-Chloro-4,5-dimethylphenol

  • MDL number MFCD00002173
  • Cas Number : 1124-04-5
  • Molecular Weight : 156.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9104

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9104
Availability details for
Quantity
  • Availability
  • Add to Cart