COM448653046 Combi-Blocks, Inc.

Allantoic acid

  • MDL number MFCD00007949
  • Cas Number : 99-16-1
  • Molecular Weight : 176.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9358

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9358
Availability details for
Quantity
  • Availability
  • Add to Cart