COM448653129 Combi-Blocks, Inc.

4-[(1-Azetidinyl)methyl]phenylboronic acid pinacol ester

  • MDL number MFCD18735839
  • Cas Number : 1036990-23-4
  • Molecular Weight : 273.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9448

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9448
Availability details for
Quantity
  • Availability
  • Add to Cart