COM448653244 Combi-Blocks, Inc.

(Imidazo[1,2-a]pyridin-5-yl)acetylene

  • MDL number MFCD22199325
  • Cas Number : 1374115-61-3
  • Molecular Weight : 142.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9570

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9570
Availability details for
Quantity
  • Availability
  • Add to Cart