COM448653452 Combi-Blocks, Inc.

2-(Boc-amino)-5-bromoisonicotinaldehyde

  • MDL number MFCD18249889
  • Cas Number : 1260667-46-6
  • Molecular Weight : 301.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9796

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QB-9796
Availability details for
Quantity
  • Availability
  • Add to Cart