COM448653640 Combi-Blocks, Inc.

1,1,1,3-Tetrafluorobutane

  • MDL number MFCD07777212
  • Cas Number : 86884-13-1
  • Molecular Weight : 130.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0005

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0005
Availability details for
Quantity
  • Availability
  • Add to Cart