COM448653657 Combi-Blocks, Inc.

2-Bromobenzo[d]thiazol-6-amine

  • MDL number MFCD11977496
  • Cas Number : 945400-80-6
  • Molecular Weight : 229.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0023

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0023
Availability details for
Quantity
  • Availability
  • Add to Cart