COM448653658 Combi-Blocks, Inc.

N-1-Boc-2-(1',3'-dihydro-1',3'-dioxo-2'h-isoindol-2'-yl)ethylamine

  • MDL number MFCD08059580
  • Cas Number : 77361-32-1
  • Molecular Weight : 290.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0024

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0024
Availability details for
Quantity
  • Availability
  • Add to Cart