COM448653664 Combi-Blocks, Inc.

3,3,4,4-Tetrafluorotetrahydrofuran

  • MDL number MFCD02093328
  • Cas Number : 374-33-4
  • Molecular Weight : 144.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0031

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0031
Availability details for
Quantity
  • Availability
  • Add to Cart