COM448653667 Combi-Blocks, Inc.

Methyl 2-amino-3h-benzo[d]imidazole-5-carboxylate

  • MDL number MFCD18157726
  • Cas Number : 106429-38-3
  • Molecular Weight : 191.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0034

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0034
Availability details for
Quantity
  • Availability
  • Add to Cart