COM448653670 Combi-Blocks, Inc.

N-(2-(Benzylamino)ethyl) cyclopropanamine

  • MDL number MFCD09037845
  • Cas Number : 736908-55-7
  • Molecular Weight : 190.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0037

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0037
Availability details for
Quantity
  • Availability
  • Add to Cart