COM448653675 Combi-Blocks, Inc.

4-Bromo-2-methylphenyl isocyanate

  • MDL number MFCD00037039
  • Cas Number : 1591-98-6
  • Molecular Weight : 212.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0042

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0042
Availability details for
Quantity
  • Availability
  • Add to Cart