COM448653676 Combi-Blocks, Inc.

Ethyl 6-chloro-1h-pyrrolo[2,3-b]pyridine-2-carboxylate

  • MDL number MFCD09751752
  • Cas Number : 287384-84-3
  • Molecular Weight : 224.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0043

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0043
Availability details for
Quantity
  • Availability
  • Add to Cart