COM448653678 Combi-Blocks, Inc.

Ethylene glycol bis(3-aminopropyl) ether

  • MDL number MFCD00059938
  • Cas Number : 2997-01-5
  • Molecular Weight : 176.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0045

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0045
Availability details for
Quantity
  • Availability
  • Add to Cart