COM448653679 Combi-Blocks, Inc.

3-Bromo-5-(trifluoromethyl)mandelic acid

  • MDL number MFCD13194211
  • Cas Number : 1214339-37-3
  • Molecular Weight : 299.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0046

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0046
Availability details for
Quantity
  • Availability
  • Add to Cart