COM448653680 Combi-Blocks, Inc.

2-((Piperidin-4-yl)methoxy)-6-(3-methoxyphenyl)pyrazine

  • MDL number MFCD14585390
  • Cas Number : 1206970-23-1
  • Molecular Weight : 299.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0047

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0047
Availability details for
Quantity
  • Availability
  • Add to Cart