COM448653688 Combi-Blocks, Inc.

(2,2-Dimethyltetrahydro-2h-pyran-4-yl)methanamine

  • MDL number MFCD10698085
  • Cas Number : 50675-25-7
  • Molecular Weight : 143.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0055

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0055
Availability details for
Quantity
  • Availability
  • Add to Cart