COM448653708 Combi-Blocks, Inc.

Methyl (2r,3s)-3-(tert-butoxycarbonylamino)-2-hydroxy-3-phenylpropionate

  • MDL number MFCD09037385
  • Cas Number : 124605-42-1
  • Molecular Weight : 295.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0075

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0075
Availability details for
Quantity
  • Availability
  • Add to Cart