COM448653733 Combi-Blocks, Inc.

4-Methoxy-1-methyl-2-oxo-1,2,5,6-tetrahydropyridine-3-carbonitrile

  • MDL number MFCD26937557
  • Cas Number : 1421314-07-9
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0104

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0104
Availability details for
Quantity
  • Availability
  • Add to Cart