COM448653740 Combi-Blocks, Inc.

2-(2-Hydroxyethylamino)-n-(2-chlorophenyl)acetamide

  • MDL number MFCD09037908
  • Cas Number : 215649-71-1
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0111

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0111
Availability details for
Quantity
  • Availability
  • Add to Cart