COM448653748 Combi-Blocks, Inc.

1-Chloro-1,2-dibromo-2-fluoropropane

  • MDL number MFCD07784196
  • Cas Number : 885276-04-0
  • Molecular Weight : 254.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0120

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0120
Availability details for
Quantity
  • Availability
  • Add to Cart