COM448653762 Combi-Blocks, Inc.

Benzoyl leuco methylene blue

  • MDL number MFCD00059153
  • Cas Number : 1249-97-4
  • Molecular Weight : 389.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0134

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0134
Availability details for
Quantity
  • Availability
  • Add to Cart