COM448653763 Combi-Blocks, Inc.

N-(2-Bromothiazol-4-yl)-2,2,2-trifluoroacetamide

  • MDL number MFCD17169924
  • Cas Number : 59134-90-6
  • Molecular Weight : 275.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0135

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0135
Availability details for
Quantity
  • Availability
  • Add to Cart