COM448653768 Combi-Blocks, Inc.

1-Cyclohexyl-piperazine dihydrochloride

  • MDL number MFCD00039024
  • Cas Number : 245487-40-5
  • Molecular Weight : 241.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0140

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0140
Availability details for
Quantity
  • Availability
  • Add to Cart