COM448653777 Combi-Blocks, Inc.

1,1,2,2-Tetrafluoro-2-(1,1,2,2-tetrafluoroethoxy)ethanesulphonyl fluoride

  • MDL number MFCD07778617
  • Cas Number : 104729-49-9
  • Molecular Weight : 300.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0149

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0149
Availability details for
Quantity
  • Availability
  • Add to Cart