COM448653778 Combi-Blocks, Inc.

2-Chloro-3-ethynylpyridine

  • MDL number MFCD13189764
  • Cas Number : 1196156-69-0
  • Molecular Weight : 137.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0150

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0150
Availability details for
Quantity
  • Availability
  • Add to Cart