COM448653781 Combi-Blocks, Inc.

4-(4-Methylphenyl)-1h-imidazole-1,2-diamine

  • MDL number MFCD03086073
  • Cas Number : 15965-80-7
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0154

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0154
Availability details for
Quantity
  • Availability
  • Add to Cart