COM448653789 Combi-Blocks, Inc.

(S)-1-(2-Fluorophenyl)ethanol

  • MDL number MFCD03092992
  • Cas Number : 171032-87-4
  • Molecular Weight : 140.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0162

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?QC-0162
Availability details for
Quantity
  • Availability
  • Add to Cart